This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01206224_g1
See other PSMG3 GE Assays ›
Gene Symbol:
Psmg3
Gene Name:
proteasome (prosome, macropain) assembly chaperone 3
Gene Aliases:
1810042K04Rik, 4930403H09Rik, AI303239
Chromosome Location:
Chr.5: 139823594 - 139827003 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01206224_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Psmg3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025604.3 NP_079880.1 1 - 2 121 93
GenBank mRNA BC058692.1 - 1 - 2 107 6818127 92

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC058692.1, NM_025604.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Psmg3

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500032105.mm.1 TC0500003743.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500032105.mm.1 NM_025604.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500032105.mm.1 NM_025604.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500032102.mm.1 TC0500003743.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500032102.mm.1 NM_025604.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500032102.mm.1 NM_025604.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0