This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01206242_g1
See other NUDT1 GE Assays ›
Gene Symbol:
Nudt1
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 1
Gene Aliases:
Mth1
Chromosome Location:
Chr.5: 140321627 - 140338137 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01206242_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008637.1 NP_032663.1 1 - 2 30 57
GenBank mRNA AK168312.1 - 1 - 2 33 57
D49956.1 - 1 - 2 30 57

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK168312.1, D49956.1, NM_008637.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nudt1

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

purine nucleotide catabolic process
dGTP catabolic process
DNA repair
DNA protection
dATP catabolic process
RNA binding
8-oxo-7,8-dihydroguanosine triphosphate pyrophosphatase activity
hydrolase activity
snoRNA binding
8-oxo-7,8-dihydrodeoxyguanosine triphosphate pyrophosphatase activity
GTP diphosphatase activity
metal ion binding
ATP diphosphatase activity
m7G(5')pppN diphosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500015526.mm.1 TC0500001689.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500015526.mm.1 NM_008637.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015526.mm.1 NM_008637.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015520.mm.1 TC0500001689.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500015520.mm.1 NM_008637.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500015520.mm.1 NM_008637.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0