Assay Details
| Gene Symbol: Fbxo24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027708.1 | NP_081984.1 | 2 - 3 | 239 | 94 | |
| GenBank mRNA | AK016870.1 | - | 2 - 3 | 240 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo24
Genomic Map