This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01207100_m1
See other FBXO24 GE Assays ›
Gene Symbol:
Fbxo24
Gene Name:
F-box protein 24
Gene Aliases:
4933422D21Rik, Fbx24
Chromosome Location:
Chr.5: 137612505 - 137625584 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01207100_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fbxo24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027708.1 NP_081984.1 5 - 6 895 64
GenBank mRNA AK016870.1 - 5 - 6 896 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003739.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500031470.mm.1 NM_027708.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500031470.mm.1 NM_027708.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500031470.mm.1 TC0500003739.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500031471.mm.1 NM_027708.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500031471.mm.1 NM_027708.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500031471.mm.1 TC0500003739.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline