Assay Details
| Gene Symbol: Fbxo24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027708.1 | NP_081984.1 | 8 - 9 | 1372 | 107 | |
| GenBank mRNA | AK016870.1 | - | 8 - 9 | 1373 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991401 | AK016870.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991401 | NM_027708.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399059 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621657 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 486143 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 496064 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713827 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111137 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AB359227.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK020445.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK037414.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK053460.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK080354.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK080810.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK090117.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK139771.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK146406.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK147712.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK159025.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK159056.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK161569.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK170452.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | AK171993.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC026940.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC040066.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC043668.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC050846.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC052459.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC052897.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC059100.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC061633.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC067391.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC068310.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC072647.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC096449.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | BC096449.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | M10062.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | M64837.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | NM_001033205.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | NM_026521.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | NM_178649.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | U70139.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | XM_001474249.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | XM_001479569.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355289 | XM_001480214.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 24400 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76520 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991401 | AK016870.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991401 | NM_027708.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05761080 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01231977 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05761058 | - | - |
| Whole Mouse Genome G4122A | A_51_P133012 | AK016870.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P133012 | NM_027708.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01231977 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1431864_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534873 | - | - |
| Mouse Genome 430 2.0 Array | 1431864_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1431864_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71176_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71176_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500031463.mm.1 | NM_027708.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031463.mm.1 | NM_027708.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031463.mm.1 | TC0500003739.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500033840.mm.1 | NM_027708.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500033840.mm.1 | NM_027708.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500033840.mm.1 | TC0500003739.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1431864_at | - | - |
| Mouse Expression Set 430 | 1431864_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431864_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1431864_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 532183 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2637347 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2601630 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2601630 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2637347 | - | - |

Fbxo24
Genomic Map