This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01207170_m1
See other ORAI2 GE Assays ›
Gene Symbol:
Orai2
Gene Name:
ORAI calcium release-activated calcium modulator 2
Gene Aliases:
A730041O15Rik, Tmem142b
Chromosome Location:
Chr.5: 136147459 - 136170656 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01207170_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Orai2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178751.3 NP_848866.2 2 - 3 358 80
GenBank mRNA AB271215.1 - 1 - 2 211 80
AK036013.1 - 2 - 3 348 80
AK042940.1 - 2 - 3 359 80
AK079619.1 - 2 - 3 304 80
AM712356.1 - 2 - 3 268 80
AM712357.1 - 1 - 2 226 80
BB640180.1 - 2 - 3 358 80
BC066070.1 - 2 - 3 305 6853893 80
BC115648.1 - 2 - 3 284 40103121 80
BC115649.1 - 2 - 3 284 40103122 80

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Orai2

Panther Classification:

Molecular Function -

ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

store-operated calcium entry
calcium ion transmembrane transport
store-operated calcium channel activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500030885.mm.1 TC0500003415.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030885.mm.1 NM_178751.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500030885.mm.1 NM_178751.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500030888.mm.1 TC0500003415.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030888.mm.1 NM_178751.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500030888.mm.1 NM_178751.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500030890.mm.1 TC0500003415.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030890.mm.1 NM_178751.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500030890.mm.1 NM_178751.3 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 414754 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0