Assay Details
| Gene Symbol: Prkg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013833.3 | NP_001013855.1 | 7 - 8 | 1099 | 76 | |
| NM_011160.3 | NP_035290.1 | 7 - 8 | 1022 | 76 | ||
| GenBank mRNA | AF084547.1 | - | 7 - 8 | 1022 | 76 | |
| AK040353.1 | - | 1 - 2 | 156 | 76 | ||
| AK083991.1 | - | 7 - 8 | 1099 | 76 | ||
| BC113162.1 | - | 7 - 8 | 1004 | 40060049 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prkg1
Genomic Map