This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01207871_m1
See other NDUFB9 GE Assays ›
Gene Symbol:
Ndufb9
Gene Name:
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9
Gene Aliases:
1190008J14Rik
Chromosome Location:
Chr.15: 58933810 - 58939489 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01207871_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufb9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023172.3 NP_075661.1 2 - 3 316 65
GenBank mRNA AK002254.1 - 2 - 3 317 65
AK003540.1 - 2 - 3 317 65
AK004522.1 - 2 - 3 317 65
BC031539.1 - 2 - 3 298 3979845 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000423.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufb9

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
transport
oxidation-reduction process
mitophagy in response to mitochondrial depolarization
molecular_function
NADH dehydrogenase (ubiquinone) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500003005.mm.1 NM_023172.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500003005.mm.1 NM_023172.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500003005.mm.1 TC1500000423.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-svqt6:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0