This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01208650_m1
See other DYDC1 GE Assays ›
Gene Symbol:
Dydc1
Gene Name:
DPY30 domain containing 1
Gene Aliases:
1700029M23Rik
Chromosome Location:
Chr.14: 41072911 - 41092197 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01208650_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dydc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027094.1 NP_081370.1 1 - 2 126 114
GenBank mRNA AK006511.1 - 1 - 2 126 114
BC049632.1 - 1 - 2 125 6771945 114

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK006511.1, BC049632.1, NM_027094.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400003557.mm.1 NM_027094.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003557.mm.1 NM_027094.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003557.mm.1 TC1400000410.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400003559.mm.1 NM_027094.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003559.mm.1 NM_027094.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003559.mm.1 TC1400000410.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400003558.mm.1 NM_027094.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003558.mm.1 NM_027094.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003558.mm.1 TC1400000410.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-vvsgg:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0