This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01208833_m1
See other PPARGC1A GE Assays ›
Gene Symbol:
Ppargc1a
Gene Name:
peroxisome proliferative activated receptor, gamma, coactivator 1 alpha
Gene Aliases:
A830037N07Rik, ENSMUSG00000079510, Gm11133, PGC-1, PGC-1v, Pgc-1alpha, Pgc1, Pgco1, Ppargc1
Chromosome Location:
Chr.5: 51454249 - 52115958 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01208833_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppargc1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008904.2 NP_032930.1 4 - 5 692 70
NR_027710.1 4 - 5 692 70
GenBank mRNA AB061324.1 - 4 - 5 615 70
AF049330.1 - 4 - 5 644 70
AK138668.1 - 4 - 5 693 70
AK143753.1 - 4 - 5 644 70
BC066868.1 - 4 - 5 587 30094033 70
JX866948.1 - 4 - 5 580 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002394.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppargc1a

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

response to dietary excess
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
mitochondrion organization
positive regulation of mitochondrion organization
response to muscle activity
negative regulation of smooth muscle cell migration
respiratory electron transport chain
positive regulation of cellular metabolic process
circadian regulation of gene expression
cellular response to oxidative stress
regulation of circadian rhythm
negative regulation of neuron apoptotic process
cellular respiration
negative regulation of glycolytic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fatty acid oxidation
rhythmic process
positive regulation of smooth muscle cell proliferation
positive regulation of sequence-specific DNA binding transcription factor activity
negative regulation of neuron death
positive regulation of cellular respiration
positive regulation of mitochondrial DNA metabolic process
positive regulation of muscle tissue development
negative regulation of receptor activity
regulation of N-methyl-D-aspartate selective glutamate receptor activity
positive regulation of energy homeostasis
positive regulation of ATP biosynthetic process
nucleotide binding
RNA polymerase II transcription cofactor activity
nucleic acid binding
DNA binding
chromatin binding
transcription coactivator activity
RNA binding
protein binding
transcription factor binding
ligand-dependent nuclear receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
chromatin DNA binding
ubiquitin protein ligase binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-s8bwd:80/100.66.79.198:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline