This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01209589_m1
See other SSR3 GE Assays ›
Gene Symbol:
Ssr3
Gene Name:
signal sequence receptor, gamma
Gene Aliases:
0610038P07Rik, AL022999, AU022074, AW553833, TRAPG
Chromosome Location:
Chr.3: 65379657 - 65392553 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01209589_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssr3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026155.3 NP_080431.1 1 - 2 146 81
GenBank mRNA AK002818.1 - 1 - 2 147 81
AK029353.1 - 1 - 2 147 81
AK076235.1 - 1 - 2 179 81
AK088555.1 - 1 - 2 151 81
AK150255.1 - 1 - 2 140 81
AK192841.1 - 1 - 2 67 81
AK194871.1 - 1 - 2 67 81
AK199053.1 - 1 - 2 67 81
AK206034.1 - 1 - 2 67 81
BC011111.1 - 1 - 2 153 3965897 81
BC020335.1 - 1 - 2 118 81
CT010172.1 - 1 - 2 130 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002094.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssr3

Gene Ontology Categories:

Function(s)

cotranslational protein targeting to membrane

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300015815.mm.1 NM_026155.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015815.mm.1 NM_026155.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300015815.mm.1 TC0300002094.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300015814.mm.1 NM_026155.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300015814.mm.1 NM_026155.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300015814.mm.1 TC0300002094.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 476192 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0