This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01209590_m1
See other SSR3 GE Assays ›
Gene Symbol:
Ssr3
Gene Name:
signal sequence receptor, gamma
Gene Aliases:
0610038P07Rik, AL022999, AU022074, AW553833, TRAPG
Chromosome Location:
Chr.3: 65379657 - 65392553 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01209590_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssr3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026155.3 NP_080431.1 3 - 4 377 113
GenBank mRNA AK002818.1 - 3 - 4 378 113
AK029353.1 - 3 - 4 378 113
AK076235.1 - 3 - 4 410 113
AK088555.1 - 3 - 4 382 113
AK150255.1 - 3 - 4 371 113
AK162235.1 - 1 - 2 81 113
BC011111.1 - 3 - 4 384 3965897 113
BC020335.1 - 3 - 4 349 113
CT010172.1 - 3 - 4 361 113

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssr3

Gene Ontology Categories:

Function(s)

cotranslational protein targeting to membrane

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0