This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01209671_m1
See other CDH4 GE Assays ›
Gene Symbol:
Cdh4
Gene Name:
cadherin 4
Gene Aliases:
AW120700, R-Cadh, R-cadherin, Rcad
Chromosome Location:
Chr.2: 179442478 - 179899377 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01209671_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009867.2 NP_033997.1 7 - 8 1192 111
GenBank mRNA D14888.1 - 7 - 8 1102 111
X69966.1 - 7 - 8 1048 111

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002772.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh4

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
axon guidance
positive regulation of axon extension
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200022623.mm.1 NM_009867.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200022623.mm.1 TC0200002772.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mfdv2:80/100.66.77.127:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0