This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01209699_m1
See other GLUD1 GE Assays ›
Gene Symbol:
Glud1
Gene Name:
glutamate dehydrogenase 1
Gene Aliases:
AI118167, Gdh-X, Glud, Gludl
Chromosome Location:
Chr.14: 34310727 - 34345033 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01209699_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glud1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008133.4 NP_032159.1 5 - 6 909 89
GenBank mRNA AK075971.1 - 5 - 6 813 89
AK089152.1 - 5 - 6 812 89
AK150637.1 - 5 - 6 909 89
AK161876.1 - 1 - 2 43 89
BC052724.1 - 5 - 6 883 6812055 89
BC057347.1 - 5 - 6 883 6812055 89
X57024.1 - 5 - 6 814 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000373.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Glud1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid metabolic process
glutamate biosynthetic process
glutamate catabolic process
glutamine metabolic process
response to aluminum ion
positive regulation of insulin secretion
oxidation-reduction process
tricarboxylic acid metabolic process
nucleotide binding
glutamate dehydrogenase (NAD+) activity
glutamate dehydrogenase [NAD(P)+] activity
protein binding
ATP binding
GTP binding
oxidoreductase activity
enzyme binding
ADP binding
NAD+ binding
leucine binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0