This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01210227_m1
See other PPP2R2C GE Assays ›
Gene Symbol:
Ppp2r2c
Gene Name:
protein phosphatase 2, regulatory subunit B, gamma
Gene Aliases:
6330548O06Rik, IMYPNO, IMYPNO1, PR52
Chromosome Location:
Chr.5: 36866843 - 36955078 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01210227_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp2r2c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172994.2 NP_766582.1 2 - 3 423 83
GenBank mRNA AK049830.1 - 2 - 3 309 83
AY348866.1 - 2 - 3 425 83
BC059811.1 - 2 - 3 423 6849535 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000417.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp2r2c

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
protein phosphatase type 2A regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500004107.mm.1 NM_172994.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500004107.mm.1 NM_172994.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500004107.mm.1 TC0500000417.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500004108.mm.1 NM_172994.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500004108.mm.1 NM_172994.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500004108.mm.1 TC0500000417.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500004106.mm.1 NM_172994.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500004106.mm.1 NM_172994.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500004106.mm.1 TC0500000417.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline