This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01210321_m1
See other ABLIM2 GE Assays ›
Gene Symbol:
Ablim2
Gene Name:
actin-binding LIM protein 2
Gene Aliases:
AI606905, C230091L11
Chromosome Location:
Chr.5: 35757855 - 35884979 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01210321_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ablim2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177696.1 NP_001171167.1 3 - 4 512 84
NM_001177697.1 NP_001171168.1 3 - 4 512 84
NM_001177698.1 NP_001171169.1 3 - 4 512 84
NM_001177699.1 NP_001171170.1 3 - 4 512 84
NM_001177700.1 NP_001171171.1 3 - 4 512 84
NM_177678.7 NP_808346.3 3 - 4 512 84
GenBank mRNA AJ748602.1 - 3 - 4 468 84
AJ748603.1 - 3 - 4 468 84
AK046243.1 - 3 - 4 463 84
AK046879.1 - 3 - 4 461 84
AK082707.1 - 3 - 4 462 84
AK154494.1 - 3 - 4 514 84
AY274116.1 - 3 - 4 439 84
BC141125.1 - 3 - 4 372 9055737 84
DQ413175.1 - 2 - 3 341 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000400.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ablim2

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
cytoskeleton organization
actin cytoskeleton organization
positive regulation of transcription from RNA polymerase II promoter
molecular_function
actin binding
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 TC0500000400.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_001177696.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_001177696.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_001177697.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_001177697.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_001177698.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_001177698.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_001177699.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_001177699.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_001177700.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_001177700.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_177678.7.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003987.mm.1 NM_177678.7 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8t6tc:80/100.66.75.151:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0