This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01210326_m1
See other ABLIM2 GE Assays ›
Gene Symbol:
Ablim2
Gene Name:
actin-binding LIM protein 2
Gene Aliases:
AI606905, C230091L11
Chromosome Location:
Chr.5: 35757855 - 35884979 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01210326_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ablim2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177696.1 NP_001171167.1 7 - 8 938 55
NM_001177697.1 NP_001171168.1 7 - 8 938 55
NM_001177698.1 NP_001171169.1 7 - 8 938 55
NM_001177699.1 NP_001171170.1 7 - 8 938 55
NM_001177700.1 NP_001171171.1 7 - 8 938 55
NM_177678.7 NP_808346.3 7 - 8 938 55
GenBank mRNA AJ748602.1 - 7 - 8 894 55
AJ748603.1 - 7 - 8 894 55
AK046243.1 - 7 - 8 889 55
AK046879.1 - 7 - 8 887 55
AK082707.1 - 7 - 8 888 55
AK154494.1 - 7 - 8 940 55
AY274116.1 - 7 - 8 865 55
BC141125.1 - 7 - 8 798 9055737 55

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000400.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ablim2

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
cytoskeleton organization
actin cytoskeleton organization
positive regulation of transcription from RNA polymerase II promoter
molecular_function
actin binding
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 TC0500000400.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_001177696.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_001177696.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_001177697.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_001177697.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_001177698.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_001177698.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_001177699.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_001177699.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_001177700.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_001177700.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_177678.7.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003994.mm.1 NM_177678.7 Refseq
Affymetrix Mouse Clariom S Array PSR0500003996.mm.1 TC0500000400.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003996.mm.1 NM_001177696.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003996.mm.1 NM_001177696.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003996.mm.1 NM_001177698.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003996.mm.1 NM_001177698.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 TC0500000400.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_001177696.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_001177696.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_001177697.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_001177697.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_001177698.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_001177698.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_001177699.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_001177699.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_001177700.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_001177700.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_177678.7.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003993.mm.1 NM_177678.7 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0