This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01210656_m1
See other HADHB GE Assays ›
Gene Symbol:
Hadhb
Gene Name:
hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme A thiolase/enoyl-Coenzyme A hydratase (trifunctional protein), beta subunit
Gene Aliases:
4930479F15Rik, Mtpb, TP-beta
Chromosome Location:
Chr.5: 30155253 - 30184593 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01210656_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hadhb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289798.1 NP_001276727.1 1 - 2 137 102
GenBank mRNA AK033462.1 - 1 - 2 72 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK033462.1, NM_001289798.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
fatty acid beta-oxidation
metabolic process
oxidation-reduction process
fatty-acyl-CoA binding
catalytic activity
3-hydroxyacyl-CoA dehydrogenase activity
acetyl-CoA C-acyltransferase activity
enoyl-CoA hydratase activity
long-chain-enoyl-CoA hydratase activity
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
transferase activity
transferase activity, transferring acyl groups
transferase activity, transferring acyl groups other than amino-acyl groups
protein complex binding
poly(A) RNA binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rw9fd:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0