Assay Details
| Gene Symbol: Cib4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028483.1 | NP_082759.1 | 2 - 3 | 151 | 69 | |
| GenBank mRNA | AK006670.1 | - | 2 - 3 | 152 | 69 | |
| BC099558.1 | - | 2 - 3 | 97 | 69 | ||
| BC118938.1 | - | 2 - 3 | 116 | 40091772 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cib4
Genomic Map