This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01211131_m1
See other CLNK GE Assays ›
Gene Symbol:
Clnk
Gene Name:
cytokine-dependent hematopoietic cell linker
Gene Aliases:
MIST
Chromosome Location:
Chr.5: 38705588 - 38876858 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01211131_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Clnk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013748.3 NP_038776.3 15 - 16 908 65
GenBank mRNA AB021220.1 - 15 - 16 1028 65
AF187819.1 - 15 - 16 908 65
AK082826.1 - 11 - 12 646 65
BC125637.1 - 15 - 16 905 40130152 65
BC125639.1 - 15 - 16 905 40130154 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002290.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Clnk

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

immune response
transmembrane receptor protein tyrosine kinase signaling pathway
intracellular signal transduction
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500021143.mm.1 NM_013748.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021143.mm.1 NM_013748.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500021143.mm.1 TC0500002290.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline