This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01211643_g1
See other GSTO1 GE Assays ›
Gene Symbol:
Gsto1
Gene Name:
glutathione S-transferase omega 1
Gene Aliases:
AA407097, AI194287, AU018802, GSTO 1-1, GSTX, SPG-R, p28
Chromosome Location:
Chr.19: 47854989 - 47864790 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01211643_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gsto1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010362.2 NP_034492.1 1 - 2 131 89
GenBank mRNA AK027922.1 - 1 - 2 81 89
AK146834.1 - 1 - 2 93 89
AK168383.1 - 1 - 2 93 89
BC085165.1 - 1 - 2 69 30359900 89
U80819.1 - 1 - 2 131 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK027922.1, AK146834.1, AK168383.1, BC085165.1, NM_010362.2, U80819.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gsto1

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
L-ascorbic acid metabolic process
L-ascorbic acid biosynthetic process
xenobiotic catabolic process
oxidation-reduction process
negative regulation of ryanodine-sensitive calcium-release channel activity
positive regulation of ryanodine-sensitive calcium-release channel activity
cellular response to arsenic-containing substance
glutathione transferase activity
oxidoreductase activity
transferase activity
glutathione dehydrogenase (ascorbate) activity
methylarsonate reductase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900006459.mm.1 NM_010362.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900006459.mm.1 TC1900000727.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900006460.mm.1 NM_010362.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900006460.mm.1 TC1900000727.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900006461.mm.1 NM_010362.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900006461.mm.1 TC1900000727.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900006462.mm.1 NM_010362.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900006462.mm.1 TC1900000727.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900006464.mm.1 NM_010362.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900006464.mm.1 TC1900000727.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 321715 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0