This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01211716_m1
See other GTF3C2 GE Assays ›
Gene Symbol:
Gtf3c2
Gene Name:
general transcription factor IIIC, polypeptide 2, beta
Gene Aliases:
1300004C11Rik, 2610510G03Rik, AI225816, AU041069, TFIIIC110, mKIAA0011
Chromosome Location:
Chr.5: 31156005 - 31180658 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01211716_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf3c2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027901.2 NP_082177.2 1 - 2 263 68
GenBank mRNA AK004895.1 - 1 - 2 41 68
AK083585.1 - 1 - 2 264 68
AK154056.1 - 1 - 2 44 68
BC043100.1 - 1 - 2 251 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK004895.1, NM_027901.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf3c2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase III promoter
DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500019868.mm.1 TC0500003724.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500019868.mm.1 NM_027901.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019868.mm.1 NM_027901.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500019862.mm.1 TC0500003724.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500019862.mm.1 NM_027901.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019862.mm.1 NM_027901.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0