This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01211952_m1
See other KAT6A GE Assays ›
Gene Symbol:
Kat6a
Gene Name:
K(lysine) acetyltransferase 6A
Gene Aliases:
1500036M03, 9930021N24Rik, MOZ, Myst3, Zfp220
Chromosome Location:
Chr.8: 22859516 - 22943262 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01211952_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kat6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081149.1 NP_001074618.1 8 - 9 1850 81
GenBank mRNA AK028058.1 - 7 - 8 1769 81
AK036885.1 - 8 - 9 1845 81
AK082463.1 - 8 - 9 1853 81
AK132474.1 - 7 - 8 1758 81
AK143408.1 - 8 - 9 1841 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000288.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

heart morphogenesis
nucleosome assembly
transcription, DNA-templated
regulation of transcription, DNA-templated
protein acetylation
positive regulation of gene expression
chromatin modification
histone acetylation
myeloid cell differentiation
somatic stem cell population maintenance
embryonic hemopoiesis
aorta morphogenesis
negative regulation of sequence-specific DNA binding transcription factor activity
histone H3 acetylation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
face morphogenesis
cellular senescence
DNA binding
chromatin binding
histone acetyltransferase activity
protein binding
transcription factor binding
zinc ion binding
acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups
transferase activity, transferring acyl groups other than amino-acyl groups
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800002158.mm.1 NM_001081149.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800002158.mm.1 NM_001081149.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800002158.mm.1 TC0800000288.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0