This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212060_m1
See other AASDH GE Assays ›
Gene Symbol:
Aasdh
Gene Name:
aminoadipate-semialdehyde dehydrogenase
Gene Aliases:
A230062G08Rik, A830035E16, Acsf4, U26
Chromosome Location:
Chr.5: 76873659 - 76905518 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212060_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aasdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_173765.3 NP_776126.1 10 - 11 1877 101
GenBank mRNA AB095954.1 - 10 - 11 1894 101
AK038779.1 - 11 - 12 1887 101
AK136849.1 - 10 - 11 1875 101
AK148405.1 - 9 - 10 1703 101
AK215879.1 - 1 - 2 161 101
AK218868.1 - 1 - 2 161 101
BC058102.1 - 10 - 11 1862 101
BC128330.1 - 9 - 10 1749 40110405 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002641.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aasdh

Panther Classification:

Molecular Function -

ligase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
biological_process
metabolic process
oxidation-reduction process
nucleotide binding
molecular_function
catalytic activity
acyl-CoA ligase activity
L-aminoadipate-semialdehyde dehydrogenase activity
succinate-CoA ligase activity
ATP binding
3-oxo-2-(2'-pentenyl)cyclopentane-1-octanoic acid CoA ligase activity
fatty acid ligase activity
oxidoreductase activity
ligase activity
3-isopropenyl-6-oxoheptanoyl-CoA synthetase activity
2-oxo-delta3-4,5,5-trimethylcyclopentenylacetyl-CoA synthetase activity
benzoyl acetate-CoA ligase activity
2,4-dichlorobenzoate-CoA ligase activity
pivalate-CoA ligase activity
cyclopropanecarboxylate-CoA ligase activity
adipate-CoA ligase activity
citronellyl-CoA ligase activity
mentha-1,3-dione-CoA ligase activity
thiophene-2-carboxylate-CoA ligase activity
2,4,4-trimethylpentanoate-CoA ligase activity
cis-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
trans-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
branched-chain acyl-CoA synthetase (ADP-forming) activity
aryl-CoA synthetase (ADP-forming) activity
3-hydroxypropionyl-CoA synthetase activity
perillic acid:CoA ligase (ADP-forming) activity
perillic acid:CoA ligase (AMP-forming) activity
(3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (ADP-forming) activity
(3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (AMP-forming) activity
pristanate-CoA ligase activity
malonyl-CoA synthetase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500023324.mm.1 TC0500002641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500023324.mm.1 NM_173765.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500023324.mm.1 NM_173765.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500023320.mm.1 TC0500002641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500023320.mm.1 NM_173765.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500023320.mm.1 NM_173765.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500023323.mm.1 TC0500002641.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500023323.mm.1 NM_173765.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500023323.mm.1 NM_173765.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0