This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212065_g1
See other AASDH GE Assays ›
Gene Symbol:
Aasdh
Gene Name:
aminoadipate-semialdehyde dehydrogenase
Gene Aliases:
A230062G08Rik, A830035E16, Acsf4, U26
Chromosome Location:
Chr.5: 76873659 - 76905518 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212065_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aasdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK038779.1 - 2 - 3 107 74

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK038779.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

ligase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0