This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212182_m1
See other TMPRSS11E GE Assays ›
Gene Symbol:
Tmprss11e
Gene Name:
transmembrane protease, serine 11e
Gene Aliases:
A030012E10, AY787860, DESC1, TTSP
Chromosome Location:
Chr.5: 86705186 - 86745807 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212182_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmprss11e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172880.2 NP_766468.1 2 - 3 187 72
GenBank mRNA AK037235.1 - 2 - 3 189 72
AK162701.1 - 2 - 3 184 72
AY787860.1 - 2 - 3 195 72
BC115432.1 - 2 - 3 181 40094164 72
BC115433.1 - 2 - 3 181 40094170 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002687.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmprss11e

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
cognition
serine-type endopeptidase activity
protein binding
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500023729.mm.1 TC0500002687.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500023729.mm.1 NM_172880.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500023729.mm.1 NM_172880.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0