This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212370_m1
See other TMEM79 GE Assays ›
Gene Symbol:
Tmem79
Gene Name:
transmembrane protein 79
Gene Aliases:
2310042N02Rik, 2310074C17Rik
Chromosome Location:
Chr.3: 88328653 - 88334433 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212370_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem79
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024246.5 NP_077208.1 1 - 2 58 58
GenBank mRNA AK009760.1 - 1 - 2 59 58
BC003309.1 - 1 - 2 113 3154669 58

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK009760.1, BC003309.1, NM_024246.5 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem79

Gene Ontology Categories:

Function(s) Process(es)

epithelial cell maturation
hair follicle morphogenesis
cuticle development
regulated exocytosis
positive regulation of epidermis development
establishment of skin barrier
cornification
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300017460.mm.1 NM_024246.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017460.mm.1 NM_024246.5 Refseq
Affymetrix Mouse Clariom S Array PSR0300017460.mm.1 TC0300002306.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300017456.mm.1 NM_024246.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017456.mm.1 NM_024246.5 Refseq
Affymetrix Mouse Clariom S Array PSR0300017456.mm.1 TC0300002306.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline