This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212388_m1
See other GABRR1 GE Assays ›
Gene Symbol:
Gabrr1
Gene Name:
gamma-aminobutyric acid (GABA) C receptor, subunit rho 1
Gene Aliases:
GABA-C
Chromosome Location:
Chr.4: 33120424 - 33163588 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212388_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gabrr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008075.2 NP_032101.3 5 - 6 669 55
GenBank mRNA AF024620.1 - 5 - 6 669 55
BI736525.1 - 5 - 6 703 55

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000280.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gabrr1

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
chloride transport
signal transduction
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
GABA receptor activity
protein domain specific binding
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400001834.mm.1 NM_008075.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001834.mm.1 NM_008075.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400001834.mm.1 TC0400000280.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400001833.mm.1 NM_008075.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001833.mm.1 NM_008075.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400001833.mm.1 TC0400000280.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline