This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212390_m1
See other GABRR1 GE Assays ›
Gene Symbol:
Gabrr1
Gene Name:
gamma-aminobutyric acid (GABA) C receptor, subunit rho 1
Gene Aliases:
GABA-C
Chromosome Location:
Chr.4: 33120424 - 33163588 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212390_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gabrr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008075.2 NP_032101.3 9 - 10 1247 101
GenBank mRNA AF024620.1 - 9 - 10 1247 101
BG286131.1 - 192 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400000280.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel 3' Most Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gabrr1

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
chloride transport
signal transduction
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
GABA receptor activity
protein domain specific binding
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177691 AF024620.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P177691 AF024620.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441005 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409368 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409324 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203412 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495562 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96946 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493628 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938402 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 34385 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 437888 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70169 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732132 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409592 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547826 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799105 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 220186 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854126 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376168 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337874 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 32983 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 85176 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139693 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121841 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124509 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177691 AF024620.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P177691 AF024620.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177691 AF024620.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P177691 AF024620.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825339 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825335 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825329 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825326 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825316 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825319 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825327 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825317 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825321 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825331 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825330 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825323 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825333 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825325 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22825337 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05196513 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P121841 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05196535 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124509 - -
Whole Mouse Genome G4122A A_51_P177691 AF024620.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P177691 AF024620.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115561 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124509 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05196535 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P121841 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03937568 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03779804 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03779805 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03779803 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03779800 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03937569 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825478 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825452 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825383 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825411 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825342 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825350 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825454 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825529 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825337 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825444 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825352 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825390 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825511 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825537 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825329 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825429 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825490 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825533 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825396 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825313 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825461 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825394 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825466 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825502 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825346 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825485 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825326 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825423 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825483 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825469 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825358 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825339 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825532 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825480 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825531 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825440 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825519 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825395 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825456 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825353 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825470 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825374 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825460 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825367 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825526 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825446 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825450 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825408 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825523 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825467 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825538 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825333 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825471 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825524 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825320 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825473 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825388 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825492 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825314 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825323 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825426 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825321 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825438 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825365 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825312 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825463 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825317 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825348 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825488 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825364 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825448 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825521 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825331 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22825475 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825342 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825339 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825325 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825473 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03735024 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825337 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03336007 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825482 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03456011 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825469 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03501547 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03779800 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03307129 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03913865 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825343 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825478 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825317 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825314 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03231324 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03336003 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825356 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03307138 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825340 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825480 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825323 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825320 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825331 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03779803 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03913864 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03913863 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825350 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825537 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825329 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825361 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825341 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825335 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825471 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03735025 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825357 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825470 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825321 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825475 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825312 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825358 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825344 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825333 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22825327 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10503874 - -
Affymetrix Mouse Gene 1.0 ST Array 10503869 - -
Affymetrix Mouse Gene 1.0 ST Array 10503873 - -
Affymetrix Mouse Gene 1.0 ST Array 10503872 - -
Affymetrix Mouse Gene 1.0 ST Array 10503875 - -
Affymetrix Mouse Gene 1.0 ST Array 10503870 - -
Affymetrix Mouse Gene 1.0 ST Array 10503868 - -
Affymetrix Mouse Gene 1.0 ST Array 10503871 - -
Affymetrix Mouse Gene 1.0 ST Array 10503867 - -
Affymetrix HT MG-430 PM Array Plate 1450300_PM_at - -
Mouse Genome 430 2.0 Array 1450300_PM_at - -
Mouse Genome 430 2.0 Array 1450300_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14408_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14408_at - -
Affymetrix Mouse Clariom S Array PSR0400001837.mm.1 NM_008075.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400001837.mm.1 NM_008075.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400001837.mm.1 TC0400000280.mm.1 MTA TC
Mouse Expression Set 430 1450300_at - -
Mouse Expression Set 430 1450300_PM_at - -
Murine Genome U74v2 Set 100675_at - -
Murine Genome U74v2 Set 100675_PM_at - -
Mouse Genome 430A 2.0 Array 1450300_PM_at - -
Mouse Genome 430A 2.0 Array 1450300_at - -
Affymetrix HT Mouse Genome 430A Array 1450300_at - -
Affymetrix HT Mouse Genome 430A Array 1450300_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 862159 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 2900497 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002900497 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1217502 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1217502 - -
mouse-6 v1.1 (Illumina) scl25571.10.1_30-S - -
Illumina Sentrix mouse-6.v1 bead array 2900497 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl25571.10.1_30-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2900497 - -
MouseRef-8 v2 Expression BeadChip ILMN_1217502 - -
MouseRef-8 v2 Expression BeadChip ILMN_2854708 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1217502 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2854708 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline