This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212552_m1
See other ALDH18A1 GE Assays ›
Gene Symbol:
Aldh18a1
Gene Name:
aldehyde dehydrogenase 18 family, member A1
Gene Aliases:
2810433K04Rik, AI429789, Pycs
Chromosome Location:
Chr.19: 40550257 - 40588463 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212552_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldh18a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019698.2 NP_062672.2 1 - 2 155 86
NM_153554.2 NP_705782.2 1 - 2 155 86
GenBank mRNA AK134148.1 - 1 - 2 115 86
AK147711.1 - 1 - 2 75 86
AK151072.1 - 1 - 2 77 86
AK168905.1 - 1 - 2 75 86
BB839822.1 - 1 - 2 152 86
BC033427.1 - 1 - 2 86 5009115 86
BY126398.1 - 1 - 2 155 86

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK134148.1, AK147711.1, AK151072.1, AK168905.1, BC033427.1, NM_019698.2, NM_153554.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldh18a1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glutamate metabolic process
proline biosynthetic process
ornithine biosynthetic process
metabolic process
cellular amino acid biosynthetic process
phosphorylation
citrulline biosynthetic process
oxidation-reduction process
nucleotide binding
catalytic activity
glutamate 5-kinase activity
glutamate-5-semialdehyde dehydrogenase activity
ATP binding
kinase activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
transferase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900013054.mm.1 NM_019698.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013054.mm.1 NM_019698.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013054.mm.1 NM_153554.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013054.mm.1 NM_153554.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013054.mm.1 TC1900001490.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013056.mm.1 NM_019698.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013056.mm.1 NM_019698.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013056.mm.1 NM_153554.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013056.mm.1 NM_153554.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013056.mm.1 TC1900001490.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013060.mm.1 NM_019698.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013060.mm.1 NM_019698.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013060.mm.1 NM_153554.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013060.mm.1 NM_153554.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013060.mm.1 TC1900001490.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013062.mm.1 NM_019698.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013062.mm.1 NM_019698.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013062.mm.1 NM_153554.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013062.mm.1 NM_153554.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013062.mm.1 TC1900001490.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 792289 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0