Assay Details
| Gene Symbol: Cdh26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291189.1 | NP_001278118.1 | 4 - 5 | 739 | 94 | |
| NM_198656.2 | NP_941058.1 | 4 - 5 | 739 | 94 | ||
| GenBank mRNA | AK132328.1 | - | 4 - 5 | 724 | 94 | |
| BC053928.1 | - | 4 - 5 | 647 | 6337144 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh26
Genomic Map