This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01212677_m1
See other CDH26 GE Assays ›
Gene Symbol:
Cdh26
Gene Name:
cadherin-like 26
Gene Aliases:
Gm1010
Chromosome Location:
Chr.2: 178430515 - 178487366 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01212677_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291189.1 NP_001278118.1 10 - 11 1878 66
NM_198656.2 NP_941058.1 10 - 11 1878 66
GenBank mRNA AK132328.1 - 10 - 11 1863 66
BC053928.1 - 10 - 11 1786 6337144 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002760.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh26

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
biological_process
calcium ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0