This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01213390_m1
See other DHX15 GE Assays ›
Gene Symbol:
Dhx15
Gene Name:
DEAH (Asp-Glu-Ala-His) box polypeptide 15
Gene Aliases:
DBP1, DEAH9, Ddx15, HRH2, mDEAH9
Chromosome Location:
Chr.5: 52150203 - 52190546 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01213390_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dhx15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042620.2 NP_001036085.1 11 - 12 2099 95
NM_007839.3 NP_031865.2 11 - 12 2099 95
NR_104311.1 11 - 12 2091 95
GenBank mRNA AF017153.1 - 11 - 12 1947 95
AK043111.1 - 1 - 2 94 95
AK145983.1 - 11 - 12 2049 95
AK146441.1 - 11 - 12 2046 95
AK151015.1 - 11 - 12 2062 95
AK151757.1 - 11 - 12 2062 95
AK167927.1 - 11 - 12 1970 95
AK168835.1 - 11 - 12 2067 95
AK169174.1 - 11 - 12 2067 95
BC003745.1 - 11 - 12 2042 95
BC100343.1 - 11 - 12 2042 30935044 95
BC144759.1 - 11 - 12 1922 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002406.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dhx15

Panther Classification:

Molecular Function -

RNA helicase

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
mRNA processing
RNA splicing
nucleotide binding
nucleic acid binding
double-stranded RNA binding
ATP-dependent RNA helicase activity
helicase activity
protein binding
ATP binding
ATP-dependent helicase activity
hydrolase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500021704.mm.1 NM_001042620.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500021704.mm.1 NM_007839.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500021704.mm.1 NR_104311.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021704.mm.1 TC0500002406.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500021702.mm.1 NM_001042620.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500021702.mm.1 TC0500002406.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500021703.mm.1 NM_001042620.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500021703.mm.1 TC0500002406.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline