This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01213690_m1
See other GRK4 GE Assays ›
Gene Symbol:
Grk4
Gene Name:
G protein-coupled receptor kinase 4
Gene Aliases:
A830025H08Rik, GRK, Gprk2l, Gprk4
Chromosome Location:
Chr.5: 34660287 - 34755305 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01213690_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Grk4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080743.1 NP_001074212.1 1 - 2 628 128
NM_019497.2 NP_062370.2 1 - 2 628 128
GenBank mRNA AK078475.1 - 1 - 2 500 128
AK132685.1 - 1 - 2 629 128
BC150691.1 - 1 - 2 152 9087602 128
BY719486.1 - 1 - 2 500 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000384.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Grk4

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

desensitization of G-protein coupled receptor protein signaling pathway
G-protein coupled receptor internalization
protein phosphorylation
signal transduction
phosphorylation
receptor internalization
termination of G-protein coupled receptor signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
rhodopsin kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500003696.mm.1 NM_001080743.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003696.mm.1 NM_001080743.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003696.mm.1 NM_019497.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003696.mm.1 NM_019497.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500003696.mm.1 TC0500000384.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003694.mm.1 NM_001080743.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003694.mm.1 NM_001080743.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003694.mm.1 NM_019497.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003694.mm.1 NM_019497.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500003694.mm.1 TC0500000384.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003695.mm.1 NM_001080743.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003695.mm.1 NM_001080743.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003695.mm.1 NM_019497.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003695.mm.1 NM_019497.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500003695.mm.1 TC0500000384.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003697.mm.1 NM_001080743.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003697.mm.1 NM_001080743.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003697.mm.1 NM_019497.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003697.mm.1 NM_019497.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500003697.mm.1 TC0500000384.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline