| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500003698.mm.1 |
NM_001080743.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003698.mm.1 |
NM_001080743.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003698.mm.1 |
NM_019497.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003698.mm.1 |
NM_019497.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003698.mm.1 |
TC0500000384.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500003700.mm.1 |
NM_001080743.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003700.mm.1 |
NM_001080743.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003700.mm.1 |
NM_019497.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003700.mm.1 |
NM_019497.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003700.mm.1 |
TC0500000384.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500003702.mm.1 |
NM_001080743.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003702.mm.1 |
NM_001080743.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500003702.mm.1 |
TC0500000384.mm.1 |
MTA TC |