This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01213692_m1
See other GRK4 GE Assays ›
Gene Symbol:
Grk4
Gene Name:
G protein-coupled receptor kinase 4
Gene Aliases:
A830025H08Rik, GRK, Gprk2l, Gprk4
Chromosome Location:
Chr.5: 34660287 - 34755305 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01213692_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Grk4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080743.1 NP_001074212.1 3 - 4 834 121
NM_019497.2 NP_062370.2 3 - 4 834 121
GenBank mRNA AK020792.1 - 3 - 4 834 121
AK132685.1 - 3 - 4 835 121
BC150691.1 - 3 - 4 358 9087602 121

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Grk4

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

desensitization of G-protein coupled receptor protein signaling pathway
G-protein coupled receptor internalization
protein phosphorylation
signal transduction
phosphorylation
receptor internalization
termination of G-protein coupled receptor signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
rhodopsin kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline