This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01214240_g1
See other GLT8D1 GE Assays ›
Gene Symbol:
Glt8d1
Gene Name:
glycosyltransferase 8 domain containing 1
Gene Aliases:
2410004H05Rik, 5430414N14Rik, AI450005
Chromosome Location:
Chr.14: 31001347 - 31012441 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01214240_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glt8d1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001165930.1 NP_001159402.1 8 - 9 879 132
NM_029626.2 NP_083902.2 8 - 9 1408 132
GenBank mRNA AK010388.1 - 8 - 9 1137 132
AK077388.1 - 8 - 9 872 132
AK217645.1 - 2 - 3 178 132
BC055709.1 - 7 - 8 1295 132
BC058731.1 - 7 - 8 1295 132
BC069873.1 - 8 - 9 958 6314675 132

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000319.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Glt8d1

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
molecular_function
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400002677.mm.1 NM_001165930.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400002677.mm.1 NM_001165930.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400002677.mm.1 NM_029626.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400002677.mm.1 NM_029626.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400002677.mm.1 TC1400000319.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400002676.mm.1 NM_001165930.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400002676.mm.1 NM_001165930.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400002676.mm.1 NM_029626.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400002676.mm.1 NM_029626.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400002676.mm.1 TC1400000319.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400002675.mm.1 NM_001165930.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400002675.mm.1 NM_001165930.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400002675.mm.1 NM_029626.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400002675.mm.1 NM_029626.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400002675.mm.1 TC1400000319.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0