This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01214971_g1
See other GOT1L1 GE Assays ›
Gene Symbol:
Got1l1
Gene Name:
glutamic-oxaloacetic transaminase 1-like 1
Gene Aliases:
1700083M11Rik
Chromosome Location:
Chr.8: 27197459 - 27223848 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01214971_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Got1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029674.1 NP_083950.1 3 - 4 471 78
GenBank mRNA AK006984.1 - 3 - 4 458 78
BC052754.1 - 3 - 4 471 6533656 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001954.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Got1l1

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid metabolic process
biological_process
biosynthetic process
catalytic activity
transaminase activity
methionine-oxo-acid transaminase activity
transferase activity
6-aminohexanoate transaminase activity
pyridoxal phosphate binding
N2-acetyl-L-lysine aminotransferase activity
acetyldiaminopimelate aminotransferase activity
branched-chain amino acid:2-keto-4-methylthiobutyrate aminotransferase activity
D-lysine transaminase activity
aspartate-2-keto-4-methylthiobutyrate transaminase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800015533.mm.1 NM_029674.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800015533.mm.1 NM_029674.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800015533.mm.1 TC0800001954.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0