This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01214976_g1
See other GOT1L1 GE Assays ›
Gene Symbol:
Got1l1
Gene Name:
glutamic-oxaloacetic transaminase 1-like 1
Gene Aliases:
1700083M11Rik
Chromosome Location:
Chr.8: 27197459 - 27223848 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01214976_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Got1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029674.1 NP_083950.1 8 - 9 1131 90
GenBank mRNA BC052754.1 - 8 - 9 1131 6533656 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001954.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Got1l1

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid metabolic process
biological_process
biosynthetic process
catalytic activity
transaminase activity
methionine-oxo-acid transaminase activity
transferase activity
6-aminohexanoate transaminase activity
pyridoxal phosphate binding
N2-acetyl-L-lysine aminotransferase activity
acetyldiaminopimelate aminotransferase activity
branched-chain amino acid:2-keto-4-methylthiobutyrate aminotransferase activity
D-lysine transaminase activity
aspartate-2-keto-4-methylthiobutyrate transaminase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25737189 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25737188 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25737190 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P461191 AK006984.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P461191 BC052754.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P461191 NM_029674.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P461191 AK006984.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P461191 BC052754.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P461191 NM_029674.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 528210 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696034 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172478 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111421 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P461191 AK006984.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P461191 BC052754.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P461191 NM_029674.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P461191 AK006984.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P461191 BC052754.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P461191 NM_029674.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737178 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737164 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737176 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737196 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737181 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737174 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737188 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737197 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737163 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737161 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737179 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737192 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737173 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737193 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737182 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737169 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737172 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737168 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737190 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737166 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737184 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25737185 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111421 - -
Whole Mouse Genome G4122A A_51_P461191 AK006984.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P461191 BC052754.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P461191 NM_029674.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P461191 AK006984.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P461191 BC052754.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P461191 NM_029674.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111421 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07255489 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07255487 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07255484 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07255490 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25737190 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25737188 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25737189 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737148 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737181 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737100 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737118 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737188 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737134 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737190 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737144 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737164 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737096 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737112 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737108 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737153 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737172 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737115 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737121 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737114 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737162 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737105 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737143 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737094 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737149 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737156 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737173 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737197 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737103 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737182 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737090 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737196 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737129 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737142 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737168 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737151 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737174 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737154 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737192 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737130 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737098 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737178 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737166 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737087 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737179 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737117 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737140 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25737184 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737188 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737142 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737149 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06894424 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737154 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737145 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07080654 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737186 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737137 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737163 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737131 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737176 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07246487 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06710703 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737172 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737193 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737143 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737203 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06832681 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737159 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737181 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737166 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737134 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737151 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737135 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737175 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737178 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737196 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06894428 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737153 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737198 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737177 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737190 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737199 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737156 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06702490 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737148 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07175676 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737171 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737161 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737184 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737168 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07138924 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737174 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737144 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737169 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06832688 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737182 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737192 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737173 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737197 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737202 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25737164 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10577970 - -
Affymetrix Mouse Gene 1.0 ST Array 10577968 - -
Affymetrix Mouse Gene 1.0 ST Array 10577969 - -
Affymetrix Mouse Gene 1.0 ST Array 10577966 - -
Affymetrix Mouse Gene 1.0 ST Array 10577965 - -
Affymetrix Mouse Gene 1.0 ST Array 10577967 - -
Affymetrix Mouse Gene 1.0 ST Array 10577971 - -
Affymetrix Mouse Gene 1.0 ST Array 10577964 - -
Affymetrix Mouse Gene 1.0 ST Array 10577972 - -
Affymetrix HT MG-430 PM Array Plate 1439308_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1430238_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10577963 - -
Mouse Genome 430 2.0 Array 1439308_at - -
Mouse Genome 430 2.0 Array 1439308_PM_at - -
Mouse Genome 430 2.0 Array 1430238_PM_at - -
Mouse Genome 430 2.0 Array 1430238_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 76615_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 76615_PM_at - -
Affymetrix Mouse Clariom S Array PSR0800015527.mm.1 NM_029674.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800015527.mm.1 NM_029674.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800015527.mm.1 TC0800001954.mm.1 MTA TC
Mouse Expression Set 430 1439308_at - -
Mouse Expression Set 430 1430238_at - -
Mouse Expression Set 430 1439308_PM_at - -
Mouse Expression Set 430 1430238_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1439308_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430238_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430238_at - -
Affymetrix HT Mouse Genome 430B Array 1439308_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 816716 - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246289 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1246289 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0