This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215098_g1
See other ARFGAP1 GE Assays ›
Gene Symbol:
Arfgap1
Gene Name:
ADP-ribosylation factor GTPase activating protein 1
Gene Aliases:
AI115377, Arf1gap
Chromosome Location:
Chr.2: 180967225 - 180982524 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215098_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arfgap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177706.1 NP_001171177.1 3 - 4 682 93
NM_001177707.1 NP_001171178.1 4 - 5 553 93
NM_001177708.1 NP_001171179.1 3 - 4 272 93
NM_001177709.1 NP_001171180.1 3 - 4 272 93
NM_145760.3 NP_665703.2 4 - 5 485 93
GenBank mRNA AJ401461.1 - 4 - 5 443 93
AK030295.1 - 4 - 5 441 93
AK030520.1 - 3 - 4 684 93
AK134402.1 - 4 - 5 495 93
AK145879.1 - 4 - 5 469 93
AK168007.1 - 4 - 5 466 93
AK168602.1 - 3 - 4 253 93
BC027794.1 - 4 - 5 492 93
BC052922.1 - 3 - 4 220 6312351 93
BC059817.1 - 3 - 4 211 5717009 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002811.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arfgap1

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
vesicle-mediated transport
regulation of endocytosis
positive regulation of GTPase activity
GTPase activator activity
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_001177706.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_001177706.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_001177707.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_001177707.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_001177708.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_001177708.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_001177709.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_001177709.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_001177710.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_001177710.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_145760.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 NM_145760.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200023027.mm.1 TC0200002811.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0