This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215153_g1
See other SLCO4A1 GE Assays ›
Gene Symbol:
Slco4a1
Gene Name:
solute carrier organic anion transporter family, member 4a1
Gene Aliases:
OATP-E, Slc21a12
Chromosome Location:
Chr.2: 180456250 - 180474853 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215153_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco4a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_148933.1 NP_683735.1 9 - 10 2121 67
GenBank mRNA AK033598.1 - 9 - 10 1941 67
BC030719.1 - 9 - 10 2121 4501966 67
BC030720.1 - 9 - 10 2070 4505508 67
BC033602.1 - 2 - 3 323 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002792.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slco4a1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
organic anion transport
thyroid hormone metabolic process
thyroid hormone transport
anion transmembrane transport
transporter activity
organic anion transmembrane transporter activity
thyroid hormone transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200022834.mm.1 NM_148933.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022834.mm.1 NM_148933.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022834.mm.1 TC0200002792.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline