This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215354_s1
See other 4933434C23RIK GE Assays ›
Gene Symbol:
4933434C23Rik
Gene Name:
RIKEN cDNA 4933434C23 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215354_s1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4933434C23Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK017052.1 - 1 59 90
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY716734.1 - 1 59 Chr. X - 117709406 - 117710207 [+] on Build GRCm38 90

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted pseudogene 14920 Gm14920 ENSMUSG00000067441,
Gm1539,
H2A.Bbd1,
OTTMUSG00000018481
-694.3kb Sense Chr. X - 117014757 - 117015104 on NCBI Build GRCm38
Downstream predicted gene 1971 Gm1971 ENSMUSG00000072994 432.2kb Anti-sense Chr. X - 118142419 - 118145825 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2337306 AK017052.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P204196 AK017052.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119077 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT MG-430 PM Array Plate 1453894_PM_at - -
Mouse Genome 430 2.0 Array 1453894_PM_at - -
Mouse Genome 430 2.0 Array 1453894_at - -
Mouse Expression Set 430 1453894_at - -
Mouse Expression Set 430 1453894_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453894_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1453894_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 688135 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102370403 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline