Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 17 - 18 | 1575 | 156 | |
| GenBank mRNA | AB008516.1 | - | 16 - 17 | 1452 | 156 | |
| AK053765.1 | - | 17 - 18 | 1679 | 156 | ||
| AK084201.1 | - | 7 - 8 | 610 | 156 | ||
| AK088273.1 | - | 16 - 17 | 1522 | 156 | ||
| AK165850.1 | - | 6 - 7 | 483 | 156 | ||
| AK166120.1 | - | 7 - 8 | 515 | 156 | ||
| AK166164.1 | - | 8 - 9 | 664 | 156 | ||
| AK220495.1 | - | 17 - 18 | 1617 | 156 | ||
| BC019173.1 | - | 17 - 18 | 1544 | 156 | ||
| BC037654.1 | - | 16 - 17 | 1477 | 156 | ||
| BC057207.1 | - | 16 - 17 | 1479 | 156 | ||
| BC145489.1 | - | 12 - 13 | 1095 | 156 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc3
Genomic Map