Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 2 - 3 | 279 | 118 | |
| GenBank mRNA | AB008516.1 | - | 1 - 2 | 156 | 118 | |
| AK053765.1 | - | 2 - 3 | 383 | 118 | ||
| AK088273.1 | - | 2 - 3 | 280 | 118 | ||
| AK089974.1 | - | 2 - 3 | 222 | 118 | ||
| AK166298.1 | - | 2 - 3 | 221 | 118 | ||
| AK220495.1 | - | 2 - 3 | 321 | 118 | ||
| BC019173.1 | - | 2 - 3 | 248 | 118 | ||
| BC057207.1 | - | 2 - 3 | 237 | 118 | ||
| BC145489.1 | - | 1 - 2 | 196 | 118 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ttc3
Genomic Map