Assay Details
| Gene Symbol: Ttc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009441.2 | NP_033467.2 | 4 - 5 | 473 | 153 | |
| GenBank mRNA | AK053765.1 | - | 4 - 5 | 577 | 153 | |
| AK088273.1 | - | 4 - 5 | 474 | 153 | ||
| AK089974.1 | - | 4 - 5 | 416 | 153 | ||
| AK166298.1 | - | 4 - 5 | 415 | 153 | ||
| AK170087.1 | - | 3 - 4 | 421 | 153 | ||
| AK220495.1 | - | 4 - 5 | 515 | 153 | ||
| BC019173.1 | - | 4 - 5 | 442 | 153 | ||
| BC037654.1 | - | 3 - 4 | 375 | 153 | ||
| BC049277.1 | - | 3 - 4 | 412 | 153 | ||
| BC057207.1 | - | 4 - 5 | 431 | 153 | ||
| BC145489.1 | - | 3 - 4 | 390 | 153 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttc3
Genomic Map