This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215735_g1
See other PITPNM1 GE Assays ›
Gene Symbol:
Pitpnm1
Gene Name:
phosphatidylinositol transfer protein, membrane-associated 1
Gene Aliases:
DRES9, Mpt-1, Nir-2, PITPnm 1, Pitpnm, R75447, Rd9, RdgB, RdgB1
Chromosome Location:
Chr.19: 4099998 - 4113965 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215735_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pitpnm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008851.4 NP_032877.1 18 - 19 3426 74
NR_075078.1 19 - 20 3120 74
GenBank mRNA AF006467.1 - 19 - 20 3001 74
AK155377.1 - 18 - 19 3899 74
BC044893.1 - 18 - 19 2931 4481550 74
BC048150.1 - 18 - 19 3080 5706382 74
Y08922.1 - 18 - 19 3427 74

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pitpnm1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
phospholipid binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900000374.mm.1 NM_008851.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900000374.mm.1 NR_075078.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000374.mm.1 TC1900000027.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900000375.mm.1 NM_008851.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900000375.mm.1 NR_075078.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000375.mm.1 TC1900000027.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline