This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01215742_g1
See other PITPNM1 GE Assays ›
Gene Symbol:
Pitpnm1
Gene Name:
phosphatidylinositol transfer protein, membrane-associated 1
Gene Aliases:
DRES9, Mpt-1, Nir-2, PITPnm 1, Pitpnm, R75447, Rd9, RdgB, RdgB1
Chromosome Location:
Chr.19: 4099998 - 4113965 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01215742_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pitpnm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008851.4 NP_032877.1 8 - 9 1796 87
NR_075078.1 9 - 10 1490 87
GenBank mRNA AF006467.1 - 9 - 10 1371 87
AK155377.1 - 9 - 10 1446 87
BC044893.1 - 8 - 9 1301 4481550 87
BC048150.1 - 8 - 9 1450 5706382 87
Y08922.1 - 8 - 9 1797 87

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pitpnm1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
phospholipid binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900000351.mm.1 NM_008851.4 Refseq
Affymetrix Mouse Clariom S Array PSR1900000351.mm.1 NR_075078.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900000351.mm.1 TC1900000027.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline