This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01216039_m1
See other CLNK GE Assays ›
Gene Symbol:
Clnk
Gene Name:
cytokine-dependent hematopoietic cell linker
Gene Aliases:
MIST
Chromosome Location:
Chr.5: 38705588 - 38876858 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01216039_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Clnk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013748.3 NP_038776.3 2 - 3 145 72
GenBank mRNA AB021220.1 - 2 - 3 265 72
AF187819.1 - 2 - 3 145 72
BC125637.1 - 2 - 3 142 40130152 72
BC125639.1 - 2 - 3 142 40130154 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB021220.1, AF187819.1, BC125637.1, BC125639.1, NM_013748.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Clnk

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

immune response
transmembrane receptor protein tyrosine kinase signaling pathway
intracellular signal transduction
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline