This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01216042_g1
See other EIF2B4 GE Assays ›
Gene Symbol:
Eif2b4
Gene Name:
eukaryotic translation initiation factor 2B, subunit 4 delta
Gene Aliases:
Eif2b
Chromosome Location:
Chr.5: 31187558 - 31193139 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01216042_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2b4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127355.1 NP_001120827.1 12 - 13 1638 64
NM_001127356.1 NP_001120828.1 11 - 12 1860 64
NM_010122.2 NP_034252.2 12 - 13 1493 64
GenBank mRNA AK154364.1 - 12 - 13 1401 64
AK158282.1 - 12 - 13 1402 64
BC021884.1 - 12 - 13 1380 4168186 64
CD768292.1 - 3 - 4 388 64
M98035.1 - 11 - 12 1480 64
M98036.1 - 12 - 13 1446 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif2b4

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

ovarian follicle development
translation
translational initiation
regulation of translation
response to heat
response to glucose
oligodendrocyte development
negative regulation of translational initiation in response to stress
myelination
response to peptide hormone
positive regulation of GTPase activity
cellular metabolic process
negative regulation of translational initiation
cellular response to stimulus
translation initiation factor activity
guanyl-nucleotide exchange factor activity
translation initiation factor binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576984 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576981 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576980 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576978 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576982 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170134 M98035.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170134 M98036.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170134 AK154364.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170134 AK158282.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170134 BC021884.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399376 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847270 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916104 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360104 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98381 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158897 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170134 M98035.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170134 M98036.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170134 AK154364.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170134 AK158282.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170134 BC021884.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P424079 CD768292.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P424079 M98035.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P424079 M98036.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P424079 AK154364.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P424079 AK158282.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576958 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576961 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576947 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576949 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576978 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576968 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576972 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576970 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576950 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576984 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576953 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576981 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576957 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576954 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576969 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576967 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576963 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576960 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576965 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576974 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576983 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23576971 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158897 - -
Whole Mouse Genome G4122A A_52_P425589 M98035.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P425589 M98036.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P425589 BU707309.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P425589 AK154364.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P425589 AK158282.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P424079 CD768292.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P424079 M98035.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P424079 M98036.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P424079 AK154364.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P424079 AK158282.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121547 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131477 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158897 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04667146 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04667165 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04667159 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04667148 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576980 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576984 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576982 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576978 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23576981 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576949 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576961 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576954 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576904 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576901 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576944 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576963 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576928 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576908 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576946 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576947 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576968 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576981 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576931 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576911 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576914 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576983 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576903 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576971 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576957 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576965 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576978 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576933 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576920 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576936 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576958 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576970 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576934 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576935 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576906 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576927 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576969 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576972 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23576895 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576965 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576978 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576947 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04264757 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576944 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576950 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576961 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576968 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576954 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04673221 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576976 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576963 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04530324 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576951 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576974 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576952 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04530326 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04530315 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04530307 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576946 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23576981 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10529121 - -
Affymetrix Mouse Gene 1.0 ST Array 10529130 - -
Affymetrix Mouse Gene 1.0 ST Array 10529127 - -
Affymetrix Mouse Gene 1.0 ST Array 10529125 - -
Affymetrix Mouse Gene 1.0 ST Array 10529132 - -
Affymetrix Mouse Gene 1.0 ST Array 10529126 - -
Affymetrix Mouse Gene 1.0 ST Array 10529122 - -
Affymetrix Mouse Gene 1.0 ST Array 10529119 - -
Affymetrix Mouse Gene 1.0 ST Array 10529124 - -
Affymetrix Mouse Gene 1.0 ST Array 10529128 - -
Affymetrix Mouse Gene 1.0 ST Array 10529123 - -
Affymetrix Mouse Gene 1.0 ST Array 10529131 - -
Affymetrix Mouse Gene 1.0 ST Array 10529120 - -
Affymetrix Mouse Gene 1.0 ST Array 10529129 - -
Affymetrix HT MG-430 PM Array Plate 1449940_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10529118 - -
Mouse Genome 430 2.0 Array 1449940_PM_a_at - -
Mouse Genome 430 2.0 Array 1449940_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 13667_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 13667_PM_at - -
Affymetrix Murine Genome 6500A Array Msa15390_at - -
Mouse Expression Set 430 1449940_PM_a_at - -
Mouse Expression Set 430 1449940_a_at - -
Murine Genome U74v2 Set 95046_s_at - -
Murine Genome U74v2 Set 95046_PM_s_at - -
Murine Genome U74v2 Set 168311_f_at - -
Murine Genome U74v2 Set 168311_PM_f_at - -
Mouse Genome 430A 2.0 Array 1449940_a_at - -
Mouse Genome 430A 2.0 Array 1449940_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1449940_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1449940_a_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 102350358 - -
Illumina mouse-6 v1.0 expression beadchip 102350358 - -
Illumina mouse-6 v1.0 expression beadchip 5270082 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005270082 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2721894 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1259854 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2721894 - -
mouse-6 v1.1 (Illumina) scl0004057.1_8-S - -
mouse-6 v1.1 (Illumina) scl26742.12.1_68-S - -
Illumina Sentrix mouse-6.v1 bead array 5270082 - -
Illumina Sentrix mouse-6.v1 bead array 102350358 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl26742.12.1_68-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 102350358 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270082 - -
MouseRef-8 v2 Expression BeadChip ILMN_2721894 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2721894 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl26742.12.1_68-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0