This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01216049_g1
See other EIF2B4 GE Assays ›
Gene Symbol:
Eif2b4
Gene Name:
eukaryotic translation initiation factor 2B, subunit 4 delta
Gene Aliases:
Eif2b
Chromosome Location:
Chr.5: 31187558 - 31193139 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01216049_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2b4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127355.1 NP_001120827.1 8 - 9 1039 89
NM_001127356.1 NP_001120828.1 7 - 8 1261 89
NM_010122.2 NP_034252.2 8 - 9 894 89
GenBank mRNA AK154364.1 - 8 - 9 802 89
AK158282.1 - 8 - 9 803 89
BC021884.1 - 8 - 9 781 4168186 89
M98035.1 - 7 - 8 881 89
M98036.1 - 8 - 9 847 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002176.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif2b4

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

ovarian follicle development
translation
translational initiation
regulation of translation
response to heat
response to glucose
oligodendrocyte development
negative regulation of translational initiation in response to stress
myelination
response to peptide hormone
positive regulation of GTPase activity
cellular metabolic process
negative regulation of translational initiation
cellular response to stimulus
translation initiation factor activity
guanyl-nucleotide exchange factor activity
translation initiation factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500019881.mm.1 TC0500002176.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500019881.mm.1 NM_001127355.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019881.mm.1 NM_001127355.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019881.mm.1 NM_001127356.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019881.mm.1 NM_001127356.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019881.mm.1 NM_010122.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500019881.mm.1 NM_010122.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0